Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

168440

Sigma-Aldrich

α,β-Dibromohydrocinnamic acid

≥99%

Synonym(s):

2,3-dibromo-3-phenylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(Br)CH(Br)CO2H
CAS Number:
Molecular Weight:
307.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder

mp

200 °C (dec.) (lit.)

functional group

bromo
carboxylic acid
phenyl

SMILES string

OC(=O)C(Br)C(Br)c1ccccc1

InChI

1S/C9H8Br2O2/c10-7(8(11)9(12)13)6-4-2-1-3-5-6/h1-5,7-8H,(H,12,13)

InChI key

FXJWTHBNVZNQQP-UHFFFAOYSA-N

General description

Dehydrobromination of α,β-dibromohydrocinnamic acid has been studied in liquid ammonia.

Application

α,β-Dibromohydrocinnamic acid (2,3-dibromo-3-phenylpropanoic acid) was used in one-pot synthesis of enynes via microwave irradiation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydrobromination of a, ?-Dibromohydrocinnamic Acid in Liquid Ammonia1.
Paulson RV and MacGregor WS.
Journal of the American Chemical Society, 73(2), 679-681 (1951)
Synthesis of (Z)-1-bromo-1-alkenes and terminal alkynes from anti-2, 3-dibromoalkanoic acids by microwave-induced reaction.
Kuang C, et al.
Tetrahedron, 61(16), 4043-4052 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service