Skip to Content
MilliporeSigma

8.14345

Pyridinium bromide-perbromide

for synthesis

Synonym(s):

Pyridinium bromide-perbromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H6Br3N
CAS Number:
Molecular Weight:
319.82
UNSPSC Code:
12352107
EC Index Number:
254-446-8
NACRES:
NA.22
MDL number:
Form:
crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

crystals

mp

132-134 °C

InChI

1S/C5H5N.3Br/c1-2-4-6-5-3-1;;;/h1-5H;;;/p+1

InChI key

GWJBDYRGIFGTOG-UHFFFAOYSA-O

Analysis Note

Assay (iodometric): ≥ 85.0 %
Identity (IR): passes test


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library