Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

166561

Sigma-Aldrich

3-Chloropropyl acetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)3Cl
CAS Number:
Molecular Weight:
136.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.429 (lit.)

bp

80-81 °C/30 mmHg (lit.)

density

1.111 g/mL at 25 °C (lit.)

functional group

chloro
ester

SMILES string

CC(=O)OCCCCl

InChI

1S/C5H9ClO2/c1-5(7)8-4-2-3-6/h2-4H2,1H3

InChI key

KPOHQIPNNIMWRL-UHFFFAOYSA-N

General description

3-Chloropropyl acetate on reaction with strong base yields trimethylene oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of Trimethylene Oxide Formation from 3-Chloropropyl Acetate.
Dittmer DC, et al.
Journal of the American Chemical Society, 79(16), 4431-4435 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service