Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163643

Sigma-Aldrich

N,N-Dimethylthioformamide

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCSN(CH3)2
CAS Number:
Molecular Weight:
89.16
Beilstein:
1737191
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.576 (lit.)

bp

58-60 °C/1 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

[H]C(=S)N(C)C

InChI

1S/C3H7NS/c1-4(2)3-5/h3H,1-2H3

InChI key

SKECXRFZFFAANN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylthioformamide undergoes desulfurization reaction with hydrosilane under photo irradiation in the presence of a methyl iron complex. It forms a deep-orange complex with anhydrous bismuth(III) trifluoromethanesulfonate.

Application

N,N-Dimethylthioformamide was used as sulfur donor solvent to investigate the coordination chemistry of lead(II).

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kozo Fukumoto et al.
Chemical communications (Cambridge, England), 48(32), 3809-3811 (2012-01-06)
Desulfurization of N,N-dimethylthioformamide (Me(2)NCHS) by hydrosilane has been achieved under photo irradiation in the presence of a methyl iron complex. The reaction sequences have been proposed, in which silyl migration from Fe to S of thioformamide triggers the cleavage of
Ingmar Persson et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(51), 18583-18592 (2016-11-20)
The coordination chemistry of d10 s2 metal ions is strongly affected by an (at least partially) occupied d10 s2 metal ion-ligand atom antibonding orbital, which may cause a void in the coordination sphere due to repulsion between the electrons in
Krzysztof Lyczko et al.
Inorganic chemistry, 43(22), 7094-7100 (2004-10-27)
At the dissolution of anhydrous bismuth(III) trifluoromethanesulfonate in N,N-dimethylthioformamide (DMTF) a deep red-orange complex, lambda(max) = 457 nm, is formed. Bismuth(III) is reduced by the solvent to a low-valent oxidation state stabilized by the sulfur-coordinating solvent DMTF. The obtained complex
Ingmar Persson et al.
Inorganic chemistry, 50(3), 1058-1072 (2011-01-14)
The coordination chemistry of lead(II) in the oxygen donor solvents water, dimethylsulfoxide (dmso, Me(2)SO), N,N-dimethylformamide (dmf), N,N-dimethylacetamide (dma), N,N'-dimethylpropyleneurea (dmpu), and 1,1,3,3-tetramethylurea (tmu), as well as in the sulfur donor solvent N,N-dimethylthioformamide (dmtf), has been investigated by extended X-ray absorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service