Skip to Content
MilliporeSigma
All Photos(1)

Documents

162507

Sigma-Aldrich

3-Fluorobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2OH
CAS Number:
Molecular Weight:
126.13
Beilstein:
2242511
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

104-105 °C/22 mmHg (lit.)

density

1.164 g/mL at 25 °C (lit.)

SMILES string

OCc1cccc(F)c1

InChI

1S/C7H7FO/c8-7-3-1-2-6(4-7)5-9/h1-4,9H,5H2

InChI key

QDHRSLFSDGCJFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Fluorobenzyl alcohol undergoes acetylation reaction with acetic anhydride catalyzed by tribromo melamine to yield 3-fluorobenzyl acetate.

Application

3-Fluorobenzyl alcohol was used as substrate to study pH dependence of aryl-alcohol oxidase activity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tribromo melamine as novel and versatile catalyst for the formylation and acetylation of alcohols.
Hajjami M, et al.
Chinese Journal of Catalysis, 35(2), 260-263 (2013)
Rianne E van Outersterp et al.
The Analyst, 145(18), 6162-6170 (2020-09-15)
Distinguishing positional isomers, such as compounds having different substitution patterns on an aromatic ring, presents a significant challenge for mass spectrometric analyses and is a frequently encountered difficulty in, for example, drug metabolism research. In contrast to mass spectrometry, IR
Patricia Ferreira et al.
The Journal of biological chemistry, 284(37), 24840-24847 (2009-07-04)
Aryl-alcohol oxidase (AAO) is a FAD-containing enzyme in the GMC (glucose-methanol-choline oxidase) family of oxidoreductases. AAO participates in fungal degradation of lignin, a process of high ecological and biotechnological relevance, by providing the hydrogen peroxide required by ligninolytic peroxidases. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service