Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

159506

Sigma-Aldrich

4-Tritylaniline

97%

Synonym(s):

4-Triphenylmethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CC6H4NH2
CAS Number:
Molecular Weight:
335.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

255-257 °C (lit.)

SMILES string

Nc1ccc(cc1)C(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C25H21N/c26-24-18-16-23(17-19-24)25(20-10-4-1-5-11-20,21-12-6-2-7-13-21)22-14-8-3-9-15-22/h1-19H,26H2

InChI key

XYHDHXBLSLSXSR-UHFFFAOYSA-N

General description

4-Tritylaniline is also referred as p-tritylaniline.

Application

4-Tritylaniline was used in the preparation of:
  • 5-tritylisatin
  • 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate
  • bis-4,4-[6-chloro-N-(4-tritylphenyl)-[1,3,5]triazine-2,4-diamine]-[2,2-disulfonic acid]-stilbene, novel fluorophore, with potential application as an optical brightener

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larry L Klein et al.
Tetrahedron letters, 54(8), 1008-1011 (2013-03-08)
Isatins are valuable intermediates for heterocyclic chemistry. Most of the common methods for their production are less than adequate when the number and lipophilicity of substituents on the targeted isatin are increased. Our group desired such molecules and identified an
Synthesis of a Fluorophore with Improved Optical Brightness.
Okuom M, et al.
International Journal of Organic Chemistry (2013)
2, 7-Di-tert-butyl-9, 9-dimethyl-4, 5-bis (4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
Vysotsky MO and Schollmeyer D.
Acta Crystallographica Section E, Structure Reports Online, 63(1), o169-o171 (2006)
Salvatore Debonis et al.
Journal of medicinal chemistry, 51(5), 1115-1125 (2008-02-13)
The human kinesin Eg5 is a potential drug target for cancer chemotherapy. Eg5 specific inhibitors cause cells to block in mitosis with a characteristic monoastral spindle phenotype. Prolonged metaphase block eventually leads to apoptotic cell death. S-trityl-L-cysteine (STLC) is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service