Skip to Content
MilliporeSigma
All Photos(1)

Documents

159352

Sigma-Aldrich

2-Fluorophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NCO
CAS Number:
Molecular Weight:
137.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5144 (lit.)

density

1.222 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccccc1N=C=O

InChI

1S/C7H4FNO/c8-6-3-1-2-4-7(6)9-5-10/h1-4H

InChI key

VZNCSZQPNIEEMN-UHFFFAOYSA-N

General description

2-Fluorophenyl isocyanate is the main component of essential oil from Rhododendron thymifolium Maxim. Molecular geometry of 2-fluorophenyl isocyanate has been investigated by microwave spectroscopy and nuclear magnetic resonance spectroscopy.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the Essential Constituent of Rhododendron thymifolium Maxim.
Ji Z, et al.
Acta Horticulturae Sinica, 29(4), 386-388 (2002)
Microwave spectroscopy and nuclear magnetic resonance study of the molecular geometry of 2-fluorophenylisocyanate.
Masson MA, et al.
Journal of Molecular Structure, 68, 307-314 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service