Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

159026

Sigma-Aldrich

β-D-Ribofuranose 1,2,3,5-tetraacetate

98%

Synonym(s):

1,2,3,5-Tetra-O-acetyl-β-D-ribofuranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O9
CAS Number:
Molecular Weight:
318.28
Beilstein:
94078
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]26/D −11.4°, c = 10 in chloroform

mp

81-83 °C (lit.)

SMILES string

CC(=O)OC[C@H]1O[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C13H18O9/c1-6(14)18-5-10-11(19-7(2)15)12(20-8(3)16)13(22-10)21-9(4)17/h10-13H,5H2,1-4H3/t10-,11-,12-,13-/m1/s1

InChI key

IHNHAHWGVLXCCI-FDYHWXHSSA-N

Application

Starting material for nucleoside synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service