Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

158763

Sigma-Aldrich

Methyl diethylphosphonoacetate

97%

Synonym(s):

Methyl P,P-diethylphosphonoacetate, (Methoxycarbonylmethyl) diethoxyphosphine oxide, NSC 147757, [(Methoxycarbonyl)methyl]phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2CH3
CAS Number:
Molecular Weight:
210.16
Beilstein:
1782397
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.433 (lit.)

bp

127-131 °C/9 mmHg (lit.)

density

1.145 g/mL at 25 °C (lit.)

functional group

ester
phosphonate

SMILES string

CCOP(=O)(CC(=O)OC)OCC

InChI

1S/C7H15O5P/c1-4-11-13(9,12-5-2)6-7(8)10-3/h4-6H2,1-3H3

InChI key

CTSAXXHOGZNKJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for:
  • Synthesis of branches allylic fluorides
  • Regioselective Diels-Alder reactions to yield antibiotic building blocks
  • Substituted thiophenes and furans synthesis for treatment of type 2 diabetes
  • Stereoselective Diels-Alder cycloaddition reactions
  • Synthesis of pyridone alkaloids with neuritogenic activity
  • Modification of botulinum neurotoxin serotype A protease inhibitors
  • Synthesis of immunosuppressive cyclopentanediol derivatives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service