Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152803

Sigma-Aldrich

2,4-Dichlorophenylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHNH2·HCl
CAS Number:
Molecular Weight:
213.49
Beilstein:
3708828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

220-224 °C (lit.)

functional group

chloro
hydrazine

SMILES string

Cl.NNc1ccc(Cl)cc1Cl

InChI

1S/C6H6Cl2N2.ClH/c7-4-1-2-6(10-9)5(8)3-4;/h1-3,10H,9H2;1H

InChI key

DDWYGJVFURAIJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorophenylhydrazine hydrochloride was used in the synthesis of pyrazole analogues of curcumin[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyendra Mishra et al.
Bioorganic & medicinal chemistry, 16(6), 2894-2902 (2008-01-16)
Curcumin, a major yellow pigment and active component of turmeric, has been shown to possess anti-inflammatory and anti-cancer activities. Recent studies have indicated that curcumin inhibits chloroquine-sensitive (CQ-S) and chloroquine-resistant (CQ-R) Plasmodium falciparum growth in culture with an IC(50) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service