Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

152250

Sigma-Aldrich

2,4-Dichlorobenzotrifluoride

98%

Synonym(s):

2,4-Dichloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CF3
CAS Number:
Molecular Weight:
215.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

117-118 °C (lit.)

density

1.484 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(Cl)cc1Cl

InChI

1S/C7H3Cl2F3/c8-4-1-2-5(6(9)3-4)7(10,11)12/h1-3H

InChI key

KALSHRGEFLVFHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorobenzotrifluoride (2,4-Dichloro-α,α,α-trifluorotoluene ) was one of the compounds present in Niagara River fish identified by gas-liquid chromatographic-mass spectrometry.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Yurawecz
Journal - Association of Official Analytical Chemists, 62(1), 36-40 (1979-01-01)
Several unknown halogenated compounds were detected in Niagara River fish using a method similar to the AOAC multiresidue method for chlorinated pesticides in high-moisture foods. From gas-liquid chromatographic-mass spectrometric (GLC/MS) data and GLC retention times on 3 columns, 7 of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service