Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152145

Sigma-Aldrich

1,1-Diphenylethanol

98%

Synonym(s):

α-Methylbenzhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H5)2OH
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

77-81 °C (lit.)

SMILES string

CC(O)(c1ccccc1)c2ccccc2

InChI

1S/C14H14O/c1-14(15,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11,15H,1H3

InChI key

GIMDPFBLSKQRNP-UHFFFAOYSA-N

General description

The H-bonds in 1,1-diphenylethanol were studied by means of infrared (IR) absorption spectra.

Application

1,1-Diphenylethanol was used to study the ESR spectra of the spiro-cyclohexadienyl intermediate in the alkoxyl radicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K U Ingold et al.
The Journal of organic chemistry, 71(26), 9906-9908 (2006-12-16)
Grossi and Strazzari have reported (J. Org. Chem. 2000, 65, 2748-2754) that the ceric ammonium nitrate modulated photooxidation of triphenylmethanol and 1,1-diphenylethanol yielded ESR spectra of the putative spiro-cyclohexadienyl intermediates in the O-neophyl rearrangements of the corresponding alkoxyl radicals, Ph2(R)CO*
B Y Sultanli et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(6), 1121-1126 (2005-03-03)
The parameters of stretching vibrations nu(OH) (frequencies and integral intensities) have been calculated from IR spectra of a large number of H-complexes of 1,1-diphenylethanol [(C(6)H(5))(2)C(CH(3))OH] with solvents of various proton accepting strength which were not regularly reported. The data has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service