Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

143073

Sigma-Aldrich

Diglycolic acid

98%

Synonym(s):

2,2′-Oxydiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(CH2COOH)2
CAS Number:
Molecular Weight:
134.09
Beilstein:
1072117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

140-144 °C (lit.)

solubility

ethanol: soluble 5%, clear to slightly hazy, colorless to greenish-yellow

functional group

carboxylic acid
ether

SMILES string

OC(=O)COCC(O)=O

InChI

1S/C4H6O5/c5-3(6)1-9-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

QEVGZEDELICMKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diglycolic acid is a toxic metabolite of diethylene glycol (DEG) responsible for DEG-induced proximal tubular necrosis.

Application

Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions.

Biochem/physiol Actions

Diglycolic acid has antithrombogenic properties and induces vasodilation. It inhibits succinate dehydrogenase activity in human proximal tubule cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahraa Youssef et al.
Photodiagnosis and photodynamic therapy, 22, 115-126 (2018-03-28)
In this study, light-sensitive photosensitizers (Chlorin e6, Ce6) were linked to TiO2 and SiO2 nanoparticles (NPs) in order to develop new kinds of NP-based drug delivery systems for cancer treatment by PDT. TiO2 or SiO2 NPs were modified either by
Ming-Ming Yu et al.
Journal of pharmaceutical and biomedical analysis, 85, 21-27 (2013-07-23)
Controlling and minimizing the adverse effects of drugs are the key issues in ensuring the safety of drug therapy. Carboxymethyl chitosan has been widely used as an anti-adhesion material. However, recently in China there have been several reported instances of
Preparation of needle-like aragonite particles from calcium nitrate solution.
Fellner P, et al.
Acta Chimica Slovenica, 4(2), 3-10 (2011)
Miriam E Mossoba et al.
Toxicology in vitro : an international journal published in association with BIBRA, 52, 87-93 (2018-06-12)
Carboxymethyl starches are added to food products for thickening or tablet binding/filling purposes. Although they lack toxicity, their synthesis creates the chemical byproduct diglycolic acid (DGA), which is difficult to eliminate and whose toxicity is in question. A rare case
Greg M Landry et al.
Toxicology letters, 221(3), 176-184 (2013-07-06)
Diethylene glycol (DEG) is a solvent used in consumer products allowing the increased risk for consumer exposure. DEG metabolism produces two primary metabolites, 2-hydroxyethoxyacetic acid (2-HEAA) and diglycolic acid (DGA). DGA has been shown to be the toxic metabolite responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service