Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

143049

Sigma-Aldrich

Aldrithiol-2

98%

Synonym(s):

2,2′-Dithiodipyridine, 2,2′-Dipyridyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2S2
CAS Number:
Molecular Weight:
220.31
Beilstein:
154629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

56-58 °C (lit.)

solubility

methanol: soluble 50 mg/mL, clear to very slightly hazy, colorless to faintly brownish-yellow

functional group

disulfide

storage temp.

2-8°C

SMILES string

S(Sc1ccccn1)c2ccccn2

InChI

1S/C10H8N2S2/c1-3-7-11-9(5-1)13-14-10-6-2-4-8-12-10/h1-8H

InChI key

HAXFWIACAGNFHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldrithiol-2 is a nucleocapsid zinc finger targeting compound and can abrogate the infectivity of human immunodeficiency virus type-1 virions. It is a thiol reagent.

Application

Aldrithiol-2 was used in the synthesis of adenosine-5′-phosphoimidazolide. It was employed with PPh3 as a condensing agent in the amidation of carboxylic acid enantiomers for HPLC separation.
Thiol reagent. Employed with PPh3 as a condensing agent in the amidation of carboxylic acid enantiomers for HPLC separation.

Legal Information

Aldrithiol is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 1267-1267 (1994)
Journal of Chromatography A, 662, 87-87 (1994)
J L Rossio et al.
Journal of virology, 72(10), 7992-8001 (1998-09-12)
Whole inactivated viral particles have been successfully used as vaccines for some viruses, but procedures historically used for inactivation can denature virion proteins. Results have been inconsistent, with enhancement of disease rather than protection seen in some notable instances following
Analytical Sciences, 10, 17-17 (1994)
Karlhans F Che et al.
European journal of immunology, 40(8), 2248-2258 (2010-05-11)
Priming of T cells in lymphoid tissues of HIV-infected individuals occurs in the presence of HIV-1. DC in this milieu activate T cells and disseminate HIV-1 to newly activated T cells, the outcome of which may have serious implications in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service