Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137235

Sigma-Aldrich

3′,4′-Dimethylacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3COCH3
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

243 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccc(C)c(C)c1

InChI

1S/C10H12O/c1-7-4-5-10(9(3)11)6-8(7)2/h4-6H,1-3H3

InChI key

WPRAXAOJIODQJR-UHFFFAOYSA-N

Application

3′,4′-Dimethylacetophenone was used as starting reagent in the synthesis of 4-[5-(3,4-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenecarboxyamide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiuxiang Zhu et al.
Journal of the National Cancer Institute, 94(23), 1745-1757 (2002-12-05)
The cyclooxygenase-2 (COX-2) inhibitor celecoxib is thought to act as a chemopreventive agent by sensitizing cancer cells to apoptotic signals. Other COX-2 inhibitors, such as rofecoxib, are two orders of magnitude less potent than celecoxib at inducing apoptosis. The molecular
Shakila K Evans et al.
Chemical biology & drug design, 75(3), 333-337 (2010-07-28)
Acetophenones were screened for activity against positive phototaxis of Chlamydomonas cells, a process that requires co-ordinated flagellar motility. The structure-activity relationships of a series of acetophenones are reported, including acetophenones that affect flagellar motility and cell viability. Notably, 4-methoxyacetophenone, 3,4-dimethoxyacetophenone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service