Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

160520

Sigma-Aldrich

3′,4′-Dichloroacetophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3COCH3
CAS Number:
Molecular Weight:
189.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

135 °C/12 mmHg (lit.)

mp

72-74 °C (lit.)

functional group

chloro

SMILES string

CC(=O)c1ccc(Cl)c(Cl)c1

InChI

1S/C8H6Cl2O/c1-5(11)6-2-3-7(9)8(10)4-6/h2-4H,1H3

InChI key

WBPAOUHWPONFEQ-UHFFFAOYSA-N

Application

3′,4′-Dichloroacetophenone (3,4-Dichloroacetophenone) was used in the synthesis of substituted aryl aminothiazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamid Latif Siddiqui et al.
Chemical & pharmaceutical bulletin, 55(7), 1014-1017 (2007-07-03)
Different dimeric disulfides, having the basic skeleton of bis[2-amino-4-phenyl-5-thiazolyl] disulfides were synthesized in a straightforward manner from acetophenones. 2-Amino-4-phenyl-1,3-thiazoles were prepared by the reaction of thiourea with substituted acetophenones in the presence of iodine which were then converted to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service