Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

132527

Sigma-Aldrich

(S)-(+)-2-Amino-1-butanol

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1718930
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

optical activity

[α]20/D +10°, neat

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.4521 (lit.)

bp

172-174 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC[C@H](N)CO

InChI

1S/C4H11NO/c1-2-4(5)3-6/h4,6H,2-3,5H2,1H3/t4-/m0/s1

InChI key

JCBPETKZIGVZRE-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-2-Amino-1-butanol is a chiral amino alcohol that may be used in the synthesis of (S)-2-(6-benzylamino-9-isopropyl-9H-purin-2-ylamino) butan-1-ol, an (S)-enantiomer of roscovitine. It can also be used to synthesize homochiral 2-methylpyrroles. (S)-(+)-2-Amino-1-butanol is an intermediate for the synthesis of (S,S)-ethambutol, an antibacterial agent for treating tuberculosis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photooxygenation of homochiral 2-methylpyrrole derivatives of chiral amino alcohols: simple, selective access to chiral bicyclic lactams
Aydogan F and Demir AS
Tetrahedron Asymmetry, 15(2), 259-265 (2004)
Enantioselective synthesis of (S, S)-ethambutol using proline-catalyzed asymmetric a-aminooxylation and a-amination
Kotkar SP and Sudalai A
Tetrahedron Asymmetry, 17(11), 1738-1742 (2006)
Synthesis and configuration of the cyclin-dependent kinase inhibitor roscovitine and its enantiomer.
Wang S, et al
Tetrahedron Asymmetry, 12(20), 2891-2894 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service