Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

109835

Sigma-Aldrich

2-Methylpyridine

98%

Synonym(s):

2-Picoline, α-Picoline, 2-Methylpyridine, NSC 3409

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
Beilstein:
104581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

vapor pressure

10 mmHg ( 24.4 °C)

Assay

98%

autoignition temp.

995 °F

expl. lim.

8.6 %

refractive index

n20/D 1.500 (lit.)

bp

128-129 °C (lit.)

mp

−70 °C (lit.)

solubility

H2O: freely soluble
alcohol: miscible
diethyl ether: miscible

density

0.943 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccn1

InChI

1S/C6H7N/c1-6-4-2-3-5-7-6/h2-5H,1H3

InChI key

BSKHPKMHTQYZBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Picoline (2-Methylpyridine) was used to study the electron and proton transfer reactions of lumiflavin using Fourier transform ion cyclotron resonance mass spectrometry.
2-Picoline was used in the synthetic pathway for the production of dearomatized, allylated and C-H bond activated pyridine derivatives.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J Christian Lennox et al.
The journal of physical chemistry. B, 121(46), 10530-10542 (2017-11-14)
A polypyridyl ruthenium complex with fluorinated bipyridine ligands and a covalently bound tyrosine moiety was synthesized, and its photo-induced proton-coupled electron transfer (PCET) reactivity in acetonitrile was investigated with transient absorption spectroscopy. Using flash-quench methodology with methyl viologen as an
Phillip Jochmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(43), 12115-12122 (2011-09-10)
A facile and general synthetic pathway for the production of dearomatized, allylated, and C-H bond activated pyridine derivatives is presented. Reaction of the corresponding derivative with the previously reported reagent bis(allyl)calcium, [Ca(C(3)H(5))(2)] (1), cleanly affords the product in high yield.
Tianlan Zhang et al.
The journal of physical chemistry. A, 117(44), 11136-11141 (2013-09-07)
Examination of electron transfer and proton transfer reactions of lumiflavin and proton transfer reactions of the lumiflavin radical anion by Fourier transform ion cyclotron resonance mass spectrometry is described. From the equilibrium constant determined for electron transfer between 1,4-naphthoquinone and
Malcolm E Tessensohn et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(16), 2250-2257 (2017-06-14)
The voltammetric behavior of 2,3,5,6-tetramethyl-1,4-phenylenediamine was found to be able to differentiate the hydrogen acceptor abilities of electroinactive pyridine compounds in acetonitrile. Weak and strong hydrogen acceptors were distinguished through the onset of a third oxidation process that came about
Nikolaos Kritikos et al.
Journal of chromatography. A, 1403, 70-80 (2015-06-06)
In the current study, quantitative structure-retention relationships (QSRR) were constructed based on data obtained by a LC-(ESI)-QTOF-MS/MS method for the determination of amino acid analogues, following their derivatization via chloroformate esters. Molecules were derivatized via n-propyl chloroformate/n-propanol mediated reaction. Derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service