Skip to Content
MilliporeSigma
All Photos(2)

Documents

101842

Sigma-Aldrich

Cyclohexanemethylamine

98%

Synonym(s):

(Aminomethyl)cyclohexane, Hexahydrobenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2NH2
CAS Number:
Molecular Weight:
113.20
Beilstein:
635751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.462 (lit.)

bp

159-161 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

NCC1CCCCC1

InChI

1S/C7H15N/c8-6-7-4-2-1-3-5-7/h7H,1-6,8H2

InChI key

AVKNGPAMCBSNSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclohexanemethylamine was used in the synthesis of Schiff′s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. Cyclohexanemethylamine was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, characterization and fluorescence properties of hexanuclear zinc (II) complexes.
Roy P, et al.
Polyhedron, 28(6), 1133-1137 (2009)
I Perković et al.
European journal of medicinal chemistry, 51, 227-238 (2012-03-13)
The novel 1-acyl-4-cycloalkyl/arylsemicarbazides (5a-y) and 1-acyl-5-benzyloxy/hydroxycarbamoylcarbazides (8a-f) derived from the nonsteroidal anti-inflammatory drugs ibuprofen, fenoprofen and reduced ketoprofen were prepared, fully chemically characterized and evaluated for their cytostatic, antiviral and antioxidant activities. Compounds 5 and 8 consist of a region

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service