Merck
Sigma Peptide Mix (5 µm)
Sigma Peptide Mix (5 µm) application for HPLC
Materiales
analytical column
Referencia del producto
Descripción
Precios
Columna para HPLC Discovery® BIO Wide Pore C18
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 4.6 mm I.D., 5 μm particles (568222-U)
mobile phase
[A] (80:20), 0.1%TFA in water:0.1% TFA in acetonitrile; [B] (66:34), 0.1% TFA in water:0.1% TFA in acetonitrile
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
5 μL Sigma Peptide Mix P2693 (~25 μg ea. Peptide) (~25 μg ea. peptide)
sample
Sigma Peptide Mix (Cat. No. P2693)