Merck
HPLC Analysis of Ornithine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Ornithine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materiales
analytical column
Referencia del producto
Descripción
Precios
Columna quiral para HPLC Astec® CHIROBIOTIC® T
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (50:50, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 205 nm