Merck
V900468
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside
Vetec, reagent grade, ≥98%, powder
Sinónimos:
5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal
About This Item
Fórmula empírica (notación de Hill):
C14H15BrClNO6
Número de CAS:
Peso molecular:
408.63
Beilstein/REAXYS Number:
1402009
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
product name
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, Vetec, reagent grade, ≥98%
grade
reagent grade
product line
Vetec
assay
≥98%
form
powder
solubility
DMF: methanol (1:1): 50 mg/mL, clear, colorless to light yellow
storage temp.
−20°C
SMILES string
OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O
InChI
1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1
InChI key
OPIFSICVWOWJMJ-AEOCFKNESA-N
Application
X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
related product
Referencia del producto
Descripción
Precios
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Yibing Wang et al.
PloS one, 8(3), e59195-e59195 (2013-03-26)
Our study had three objectives: to extend the plasmid-based transformation protocol to a clinical isolate of C. trachomatis belonging to the trachoma biovar, to provide "proof of principle" that it is possible to "knock out" selected plasmid genes (retaining a
Enzo R Porrello et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 187-192 (2012-12-19)
We recently identified a brief time period during postnatal development when the mammalian heart retains significant regenerative potential after amputation of the ventricular apex. However, one major unresolved question is whether the neonatal mouse heart can also regenerate in response
Khanh-Van Tran et al.
Cell metabolism, 15(2), 222-229 (2012-02-14)
Adipose tissue expansion involves the enlargement of existing adipocytes, the formation of new cells from committed preadipocytes, and the coordinated development of the tissue vascular network. Here we find that murine endothelial cells (ECs) of classic white and brown fat
Masahide Sakabe et al.
Development (Cambridge, England), 139(2), 385-395 (2011-12-22)
The progress of molecular genetics has enabled us to identify the genes responsible for congenital heart malformations. However, recent studies suggest that congenital heart diseases are induced not only by mutations in certain genes, but also by abnormal maternal factors.
Amy McCurley et al.
Nature medicine, 18(9), 1429-1433 (2012-08-28)
Hypertension is a cardiovascular risk factor present in over two-thirds of people over age 60 in North America; elevated blood pressure correlates with increased risk of heart attack, stroke and progression to heart and kidney failure. Current therapies are insufficient