Documentos clave
RAD125
radiello Anesthetic Gases and Vapor Sampler
About This Item
Código UNSPSC:
41104008
NACRES:
NB.24
En este momento no podemos mostrarle ni los precios ni la disponibilidad
fabricante / nombre comercial
Radiello
técnicas
passive air sampling: suitable
aplicaciones
air monitoring
environmental
industrial hygiene
Descripción general
This kit was developed for sampling of nitrous oxide, isofluorane, ethrane, halothane, and sevorane in surgical theaters. Parts for one complete sampler are packed separately in one sealed and sterile bag. Pack of 10 sterile bags.
Each sterile bag contains:
  • 1 permeative body
  • 1 support plate
  • 1 vertical adapter
  • 1 adsorbing cartridge
Información legal
radiello is a trademark of Institi Clinici Scientifici Maugeri
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Francesc A Esteve-Turrillas et al.
Journal of chromatography. A, 1216(48), 8549-8556 (2009-10-27)
A new procedure for the passive sampling in air of benzene, toluene, ethylbenzene and xylene isomers (BTEX) is proposed. A low-density polyethylene layflat tube filled with a mixture of solid phases provided a high versatility tool for the sampling of
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
Artículos
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocolos
Protocol for radiello® Diffusive Air Sampling Application - Anaesthetic Gases and Vapors
Learn more about radiello air sampler calibration solutions for aldehydes and H2s. We also offer many air sampler calibration kits.
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Contenido relacionado
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.