Documentos clave
57633
Rotor Seal for Model 7125/7725 Injector
Tefzel®
About This Item
Código UNSPSC:
41115854
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Materiales
Tefzel®
Descripción general
Rotor Seal for Model 7125/7725/7725i Injector is an accessory for use with mainly Rheodyne® model 7725 and 7725i injectors for operation at pH 0-14.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.