Documentos clave
56656-U
Supelco
UltraShield Precolumn Waters Compatible Filter
Stainless steel frit - 0.5 micron
About This Item
Código UNSPSC:
41115700
En este momento no podemos mostrarle ni los precios ni la disponibilidad
volumen interno
1 μL
compatibilidad
for use with Waters HPLC columns
Descripción general
The UltraShield direct connect precolumn filter provides economical protection from harmful effects of sample and mobile phase particulates on modern UHPLC and high resolution HPLC columns. The UltraShield was engineered specifically for use in fast, high efficiency separations requiring high mobile phase velocities and ultra high pressure. It maintains efficiency of high performance columns assuring no loss of critical resolution. With its simple design, UltraShield 1010W installs into the Waters compatible analytical column in seconds and is leak tight to 15,000 psi.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)