Documentos
21017
Supelco
Female Luer Fitting for 1/8 in. Tubing
pkg of 20 ea
About This Item
UNSPSC Code:
12190000
packaging
pkg of 20 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Female Luer Fitting for 1/8 in. Tubing is an accessory for use with LpDNPH Cartridges suitable for air monitoring applications.
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente
¿Ya tiene este producto?
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 3
1 of 3