Documentos clave
Z740414
SecurTainer III specimen containers
size 60 mL
Seleccione un Tamaño
500 EA
US$ 660,00
US$ 660,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
500 EA
US$ 660,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.22
US$ 660,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
polyethylene cap
polypropylene container
envase
pack of 500 ea
fabricante / nombre comercial
Simport C577-60W
tamaño
60 mL
Descripción general
Chemically resistant and shatterproof, The SecurTainer III Specimen Containers are supplied with a leakproof screw cap, particularly important when transporting hazardous material. Ideal for transport and storage of urine, sputum and most liquids or particulate samples.
Containers are uniquely stackable, shatter resistant and are manufactured from virgin, high-clarity polypropylene. Available in many sizes from 20 to 120 ml. Containers and caps packaged separately in bags of 100. Non sterile. Functional temperature range: -90 °C to +100 °C.
Información legal
SecurTainer is a trademark of Simport
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)