Documentos
Z666467
Eppendorf® Easypet® 4421 pipetting aid
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41121500
manufacturer/tradename
Eppendorf® 4421 000.013 (European)
Eppendorf® 4421 000.013 (UK)
Eppendorf® 4421 000.013
availability
available only in EU
General description
Precise, safe and fatigue-free series pipetting using graduated and one-mark pipettes is now possible with the aid of Easypet, a light, hand-held device. The device responds exactly to finger pressure for slow or rapid aspiration, setting the menisus or emptying the pipette.
  • New valve control enables more sensitive control of the meniscus, resulting in more precise dispensing of the liquids
  • Suitable for pipettes of 0.1-100 mL
  • Smooth setting of the pumping efficiency
  • High aspirating speed: 25 mL in 5 sec.
  • One-finger control for the Flow-Out and Blo-Out functions
  • Cordless work for up to seven hours, continuing directly after recharging
  • Autoclavalbe pipette adapters and filter holders
  • Active pressure compensation prevents corrosion caused by aspirated vapour
  • Space saving wall holder
Components
  • transformer
  • wall holder
  • 2 × 0.45 μm membrane filters
Legal Information
Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG