Documentos clave
Z654477
Vilber Lourmat Electronic Ballast transilluminators
ECX-F20.M
About This Item
En este momento no podemos mostrarle ni los precios ni la disponibilidad
entrada CA/CC
110 / 230 V, 50-60 Hz
fabricante / nombre comercial
Vilber Lourmat 2131 2002 1
disponibilidad
available only in EU
mesa tamaño
20 cm × 20 cm
Descripción general
These 312 nm UV transilluminators are based on an electronic ballast developed by Vilber Lourmat. Available as 8 watt (ECX models) or 15 watt (ETX models). It′s 25,000 Hz frequency provides a supreme light stability and avoids the usual flickering effect. No cooling fan is necessary, as the electronic ballast produces no heat. All models have an adjustable safety screen and stainless steel frame.
  • Compact size
  • Reduced electrical consumption
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)