Z374601
Eppendorf® Combitips® Plus
maximum volume 1.0 mL
About This Item
Productos recomendados
material
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 0030 069.234
Eppendorf® 022266209
maximum volume
1.0 mL
General description
Snaps securely into Multipette pipette. Tips are not autoclavable.
Legal Information
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG