Documentos clave
OH30253065
Ohaus® Scout® Portable Touchscreen Balance
Model STX421, weighing capacity 420 g, resolution: 0.1 g, AC/DC input 230 V
Seleccione un Tamaño
1 EACH
US$ 580,00
US$ 580,00
PRICE WITHOUT NATIONAL TAXES
Fecha estimada de envío31 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EACH
US$ 580,00
About This Item
Código UNSPSC:
41111502
NACRES:
NB.13
US$ 580,00
PRICE WITHOUT NATIONAL TAXES
Fecha estimada de envío31 de mayo de 2025
Materiales
metal weighing plate
Características
CE compliant
envase
pack of 1 ea
fabricante / nombre comercial
Ohaus® 30253065
intervalo de medición de la capacidad de pesada
420 g, resolution: 0.1 g
weighing pan L
12 cm
Descripción general
Ideal for laboratory and industrial applications, the OHAUS Scout features a user-friendly color touchscreen and a compact stackable design. Designed to deliver superior performance with rapid stability testing and high resolution weighing results, these portable scales set a new standard in weighing applications. Their features also include superior overload protection and advanced connectivity options.
Aplicación
Weighing, piece counting,% weighing, check weighing, animal weighing / dynamic weighing, totalizing, density determination, display hold, mole weighing
Características y beneficios
Stability indicator, stackable design with storage cover included (except 1 mg models), mechanical and software overload / underload protection, low battery level indicator, automatic shutdown, auto-tare, slope calibration mass included with models up to 620g.
Información legal
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)