LID-F35630-EP
Lid for Eppendorf(R) FA-35-6-30 Rotor
not aerosol-tight, for 5430/5430R, aluminum
About This Item
Productos recomendados
material
aluminum
feature
not aerosol-tight
manufacturer/tradename
Eppendorf® 022654322 (NA)
Eppendorf® 5427715002 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with 5430 R
for use with 5430
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG