Merck
I2784
Sigma-Aldrich
INH2BP
≥98% (HPLC), solid
Sinónimos:
5-Iodo-6-amino-1,2-benzopyrone
About This Item
Fórmula empírica (notación de Hill):
C9H6INO2
Número de CAS:
Peso molecular:
287.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
assay
≥98% (HPLC)
form
solid
solubility
DMSO: soluble 31 mg/mL
H2O: insoluble
storage temp.
2-8°C
SMILES string
Nc1ccc2OC(=O)C=Cc2c1I
InChI
1S/C9H6INO2/c10-9-5-1-4-8(12)13-7(5)3-2-6(9)11/h1-4H,11H2
InChI key
WWRAFPGUBABZSD-UHFFFAOYSA-N
Biochem/physiol Actions
Inhibitor of poly (ADP-ribose) polymerase-1 (PARP-1); anti-apoptotic.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Visite la Librería de documentos
France Vaillancourt et al.
Arthritis research & therapy, 10(5), R107-R107 (2008-09-11)
4-Hydroxynonenal (HNE) is one of the most abundant and reactive aldehydes of lipid peroxidation products and exerts various effects on intracellular and extracellular signalling cascades. We have previously shown that HNE at low concentrations could be considered as an important
J G Mabley et al.
British journal of pharmacology, 133(6), 909-919 (2001-07-17)
Activation of poly(ADP-ribose) synthetase (PARS, also termed polyADP-ribose polymerase or PARP) has been proposed as a major mechanism contributing to beta-cell destruction in type I diabetes. In the present study, we have investigated the role of PARS in mediating the
M Endres et al.
European journal of pharmacology, 351(3), 377-382 (1998-08-28)
Peroxynitrite triggers DNA single-strand breakage, which activates the nuclear enzyme poly(ADP-ribose) synthetase (PARS). Activation of PARS depletes its substrate, NAD+, slowing the rate of glycolysis, electron transport, and ATP formation, resulting in cell necrosis. Here, we demonstrate that inhibition of
Raquel M Forsythe et al.
Shock (Augusta, Ga.), 17(3), 180-184 (2002-03-20)
Studies indicate that endotoxin (LPS) causes intestinal injury, increases inducible nitric oxide synthase (iNOS) activity, leads to increased NO production, and promotes bacterial translocation (BT). To investigate the mechanism by which LPS causes gut injury and to test the hypothesis
C Szabó et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(7), 3867-3872 (1998-05-09)
Peroxynitrite, a cytotoxic oxidant formed from nitric oxide (NO) and superoxide, induces DNA strand breakage, which activates the nuclear enzyme poly(ADP-ribose) synthase (PARS; EC 2.4.2.30). The cellular function of PARS was determined in fibroblast lines from PARS knockout animals (PARS-/-)