Documentos
HS206203
Pipette Stand
acrylic, to hold, 3 pipette(s), clear
About This Item
UNSPSC Code:
41121800
NACRES:
NB.01
material
acrylic
clear
feature
to hold 3 pipette(s)
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS206203
L × W × H
153 mm × 172 mm × 206.4 mm
General description
The Pipette Stand offers convenient and safe storage.
Features and Benefits
  • 6-Place Pipette Holder (1 in (25mm) Upper Slot, 0.8 in (20mm) Lower Slot)
  • Designed to hold most major pipette brands