Documentos clave
GE10600057
Amersham Hybond® P Western blotting membranes, PVDF
pore size 0.2 μm, sheet W × L 200 mm × 200 mm, pkg of 25 ea
About This Item
Código UNSPSC:
41105339
NACRES:
NB.22
En este momento no podemos mostrarle ni los precios ni la disponibilidad
envase
pkg of 25 ea
fabricante / nombre comercial
Cytiva 10600057
lámina An × L
200 mm × 200 mm
tamaño de poro
0.2 μm pore size
Descripción general
PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.
Almacenamiento y estabilidad
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Información legal
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
This page describes troubleshooting strategies in the detection of GST-tagged proteins.
Protocolos
This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.