Documentos clave
CLS3721
Corning® 96 Well Half-Area Microplate
flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 5, sterile, lid
About This Item
Código UNSPSC:
41121800
NACRES:
NB.13
Materiales:
black polystyrene
clear bottom
flat bottom clear
tamaño:
96 wells
pocillo D:
10.7 mm
pocillo volumen:
190 μL
pocillo volumen de trabajo:
25-125 μL
Materiales
black polystyrene
clear bottom
flat bottom clear
esterilidad
sterile
Características
lid
skirt
envase
bag of 5
fabricante / nombre comercial
Corning 3721
tamaño
96 wells
pocillo D
10.7 mm
pocillo volumen
190 μL
pocillo volumen de trabajo
25-125 μL
idoneidad
suitable for tissue
tipo de unión
Tissue Culture (TC)-treated surface
Descripción general
Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.
Información legal
Corning is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)