BCS-405G
CoolCell™ LX Freezing Container
for 12 x 1-2 mL cryogenic vials, green
About This Item
Código UNSPSC:
41103000
Productos recomendados
fabricante / nombre comercial
BioCision BCS-405G
diám. × Al
11.7 cm × 10.9 cm
pocillo diám.
12.7 mm
color
green
Descripción general
CoolCell LX controlled-rate alcohol-free cell freezing container provides a highly reproducible -1°C/minute freezing rate when placed in a -80°C freezer. No alcohol or fluids required. The insulative outer housing and thermo-conductive solid state core ensure consistent heat removal to all vials throughout the freezing period. CoolCell containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen - no "frosty" fingers. Numbered vial wells ensure easy sample identification. Beveled junction between base and lid and exposed vial tops in the base allow for quick and safe removal of frozen vials so they can be placed into long-term storage. Proven for use with stem cells, primary cells and cell lines.
Save money and go green!
CoolCell containers replace isopropanol-based freezing containers which can consume up to 12L per year of IPA and cost hundreds of dollars per unit per year to maintain. CoolCell containers instantly save money and eliminate hazardous solvents from the cell cryopreservation process. Calculate your cost savings here.
Holds 12 x 1.0 mL or 2.0 mL cryogenic vials. Cryogenic vials fill volume: 1.0 mL per vial. Fill empty wells with CoolCell Filler Vials.
Save money and go green!
CoolCell containers replace isopropanol-based freezing containers which can consume up to 12L per year of IPA and cost hundreds of dollars per unit per year to maintain. CoolCell containers instantly save money and eliminate hazardous solvents from the cell cryopreservation process. Calculate your cost savings here.
Holds 12 x 1.0 mL or 2.0 mL cryogenic vials. Cryogenic vials fill volume: 1.0 mL per vial. Fill empty wells with CoolCell Filler Vials.
Información legal
CoolCell is a trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)