Documentos clave
AXYP384240SQCS
Corning® Axygen® Deep Well Microplate
size 384 wells, Not Treated, working volume 240 μL, pkg of 10x5plates/cs, sterile
Sinónimos:
Corning cell culture plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear plate
square bottom clear polypropylene wells
maximum volume:
250 μL
size:
384 wells
working volume:
240 μL
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
clear plate
square bottom clear polypropylene wells
sterility
sterile
feature
Not Treated
manufacturer/tradename
Axygen Inc P-384-240SQ-C-S
Corning
packaging
pkg of 10x5plates/cs
maximum volume
250 μL
size
384 wells
working volume
240 μL
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)