Merck
209678
Sigma-Aldrich
Phosphorus, red
powder, 99% trace metals basis
About This Item
Fórmula empírica (notación de Hill):
P
Número de CAS:
Peso molecular:
30.97
Número CE:
Número MDL:
Código UNSPSC:
12141909
ID de la sustancia en PubChem:
densidad de vapor
0.02 (vs air)
presión de vapor
0.03 mmHg ( 21 °C)
1 mmHg ( 76.6 °C)
Ensayo
99% trace metals basis
Formulario
powder
temp. de autoignición
500 °F
resistividad
10 μΩ-cm, 20°C
mp
416 °C (subl.) (lit.)
densidad
2.34 g/mL at 25 °C (lit.)
cadena SMILES
[P]
InChI
1S/P
Clave InChI
OAICVXFJPJFONN-UHFFFAOYSA-N
sustituido por
Referencia del producto
Descripción
Precios
Pictogramas
Flame
Palabra de señalización
Warning
Frases de peligro
Clasificaciones de peligro
Aquatic Chronic 3 - Flam. Sol. 2
Código de clase de almacenamiento
4.1B - Flammable solid hazardous materials
Clase de riesgo para el agua (WGK)
WGK 3
Punto de inflamabilidad (°F)
Not applicable
Punto de inflamabilidad (°C)
Not applicable
Equipo de protección personal
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Aloysious Aravinthan et al.
Experimental gerontology, 60, 37-45 (2014-09-23)
Hepatocyte senescence is associated closely with fibrosis stage and an adverse outcome in chronic liver disease, but it is uncertain whether there is a causal relation with clinical manifestations of chronic liver disease, which was the subject of this study
Manuel Delgado-Baquerizo et al.
Nature, 502(7473), 672-676 (2013-11-01)
The biogeochemical cycles of carbon (C), nitrogen (N) and phosphorus (P) are interlinked by primary production, respiration and decomposition in terrestrial ecosystems. It has been suggested that the C, N and P cycles could become uncoupled under rapid climate change
Jacques C Finlay et al.
Science (New York, N.Y.), 342(6155), 247-250 (2013-10-12)
Human activities have increased the availability of reactive nitrogen in many ecosystems, leading to negative impacts on human health, biodiversity, and water quality. Freshwater ecosystems, including lakes, streams, and wetlands, are a large global sink for reactive nitrogen, but factors
Yuki Fujita et al.
Nature, 505(7481), 82-86 (2013-11-19)
Plant species diversity in Eurasian wetlands and grasslands depends not only on productivity but also on the relative availability of nutrients, particularly of nitrogen and phosphorus. Here we show that the impacts of nitrogen:phosphorus stoichiometry on plant species richness can
Bon-Chul Koo et al.
Science (New York, N.Y.), 342(6164), 1346-1348 (2013-12-18)
Phosphorus ((31)P), which is essential for life, is thought to be synthesized in massive stars and dispersed into interstellar space when these stars explode as supernovae (SNe). Here, we report on near-infrared spectroscopic observations of the young SN remnant Cassiopeia