20715
Hamilton® GASTIGHT® syringe, 1700 series, luer tip
1705LT, volume 50 μL, without needle, Luer Tip, needle size (not included)
About This Item
Productos recomendados
material
glass body
needle size
(not included)
feature
without needle Luer Tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80901
needle L
, n/a
volume
50 μL
Categorías relacionadas
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificados de análisis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente