Merck
V-022
Supelco
Vigabatrin solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
About This Item
Fórmula empírica (notación de Hill):
C6H11NO2
Peso molecular:
129.16
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
NC(CCC(O)=O)C=C
InChI
1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)
InChI key
PJDFLNIOAUIZSL-UHFFFAOYSA-N
Gene Information
human ... ABAT(18)
General description
Vigabatrin is a newer antiepileptic sold as Sabril® in the US, Canada, Mexico and the United Kingdom and as Sabrilex in Denmark. The drug is used as an adjunctive treatment of complex partial seizures in adults and for infantile spasms in pediatric patients one month to two years of age. This certified Snap-N-Spike® Solution standard is suitable for use in LC/MS or GC/MS applications including forensic analysis, clinical toxicology, urine drug testing, or pharmaceutical research.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Sabril is a registered trademark of Lundbeck LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Visite la Librería de documentos
The potential value of a negative finding: an illustrative example.
Kenneth Silverman et al.
JAMA psychiatry, 70(6), 571-572 (2013-04-12)
Peter Weber et al.
Epilepsy & behavior : E&B, 24(1), 138-140 (2012-04-17)
We report the case of an adolescent girl who suffered from symptomatic refractory focal epilepsy after an arteria cerebri media insult 15 years prior to this report. Five weeks after initiation of an add-on therapy with vigabatrin, she was seizure
O Heinzlef et al.
Revue neurologique, 168 Suppl 3, S62-S68 (2012-06-29)
The medicinal treatment of spasticity includes use of oral treatments (baclofène and tizanidine), botulinum toxin, intrathecal baclofène and local application of alcohol or phenol. However, spasticity may not be uncomfortable and may even be useful. Therefore, all spastic diseases do
Lisa M Clayton et al.
Ophthalmology, 119(10), 2152-2160 (2012-08-03)
To explore the relationship of peripapillary retinal nerve fiber layer (ppRNFL) thinning in individuals exposed to the antiepileptic drug vigabatrin with respect to 2 separate variables: cumulative vigabatrin exposure and severity of vigabatrin-associated visual field loss (VAVFL). Cross-sectional observational study.
Jin Yang et al.
PloS one, 7(8), e43889-e43889 (2012-09-13)
Vigabatrin (VGB) is a commonly prescribed antiepileptic drug designed to inhibit GABA-transaminase, effectively halting seizures. Unfortunately, VGB treatment is also associated with the highest frequencies of peripheral visual field constriction of any of the antiepileptic drugs and the mechanisms that