Documentos clave
Z743163
Julabo® bath covers
flat cover to fit CORIO C-B13/17 and CD-B13 /17
Seleccione un Tamaño
1 EA
US$ 432,00
US$ 432,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 432,00
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
US$ 432,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
stainless steel
envase
pack of 1 ea
fabricante / nombre comercial
Julabo 9970290
Categorías relacionadas
Descripción general
Stainless Steel Flat bath cover for 13L and 17L bath tanks. The use of a bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath. Suitable for use with Julabo® CORIO C-B13, C-B17, CD-B13, CD-B17.
Características y beneficios
  • reduces evaporation
  • prevents contamination of samples
Información legal
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)