Documentos clave
Z742941
Julabo® hightech bridge mounted circulator
Model SE-Z, temperature 20-300 °C, AC/DC input 230 V AC, US 3-pin plug (6-20P)
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
En este momento no podemos mostrarle ni los precios ni la disponibilidad
envase
pack of 1 ea
fabricante / nombre comercial
Julabo 9252218
intervalo de medición de la temperatura
20-300 °C
Descripción general
JULABO bridge mounted circulator with extendable stainless steel bridge. The adjustable bridge permits use of the circulator with a wide variety of bath tanks.
This model is suitable for both internal and external temperature control tasks. It has the power to control the temperature of even large bath tanks up to 100 L.
An integrated cooling coil enables temperature control below or near the ambient temperature.
Información legal
Julabo is a registered trademark of Julabo USA, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)