Documentos clave
Z691542
MagLab storage system
MagPette only
Seleccione un Tamaño
1 EA
US$ 50,60
US$ 50,60
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 50,60
About This Item
Código UNSPSC:
41122800
NACRES:
NB.35
US$ 50,60
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
holds 2 x pipettors
fabricante / nombre comercial
Heathrow Scientific HD23503
L × An × Al
131 mm × 112 mm × 76 mm
color
purple
Descripción general
With lab bench top space at a premium, common lab products such as pipettors, test tubes and miscellanous accessories can be easily organized with Heathrow Scientific′s magnetic lab
storage system, the MagLab. MagLab includes four magnetic pieces: “MagRack” that holds (9) 15ml and (4) 50ml tubes; “Mag Wipe” that holds a standard size (11cm x 21cm) Kimwipes; box; “MagPette” that
holds two pipettors (universal design fits most common brands); and “MagBox” that holds miscellaneous items such as pens/pencils, notes, etc. Each rack is made of robust lightweight ABS plastic in bright and
fun translucent colors and features four powerful magnets for a secure hold onto most metal surfaces or create your own holding point with the metal plate included for each segment.
Información legal
MagLab is a trademark of Heathrow Scientific
MagPette is a trademark of Heathrow Scientific
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)