Documentos clave
Z544817
Sigma-Aldrich
Aldrich® high-efficiency variable take off head
joint: ST/NS 24/40, thermometer joint: ST/NS 10/18
Seleccione un Tamaño
1 EA
US$ 1.880,00
US$ 1.880,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 1.880,00
About This Item
Código UNSPSC:
41104812
NACRES:
NB.43
US$ 1.880,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Junta
thermometer joint: ST/NS10/18
joint: ST/NS24/40
Categorías relacionadas
Descripción general
Vapor from the distillation column passes through the upper holes in the take-off head and into the condenser. The condensed liquid can be drained into a side storage flask (PTFE valve open), or allowed to return to the distillation column (PTFE valve closed) through a slot in the lower part of the head. The upper and lower ST joints permit the use of take-off head with different distillation columns and condensers. Features a precision PTFE valve, and vacuum/purge hose connections with a PTFE stopcock.
A 50mm immersion length thermometer is recommended.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)