Documentos clave
Z530271
Sigma-Aldrich
Aldrich® straight vacuum-distilling adapter
joint: ST/NS 14/20
Seleccione un Tamaño
1 EA
US$ 103,00
US$ 103,00
Fecha estimada de envío28 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 103,00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
US$ 103,00
Fecha estimada de envío28 de mayo de 2025
Junta
joint: ST/NS14/20
Descripción general
With side hose connection and drip-tip. Improves efficiency of evaporation of solvents under reduced pressure when placed between the condenser of a rotary evaporator and a chilled receiving flask. Plug the condenser vacuum take-off exit opposite the receiving flask to eliminate path competition and direct all distillate to the chilled collection flask. SJ 35/25 adapter fits standard Büchi rotary evaporator glassware.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)