Documentos clave
Z406546
Stankovic transfer adapter
joint: ST/NS 24/29, thread size 20-400
Seleccione un Tamaño
1 EA
US$ 92,60
US$ 92,60
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 92,60
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
US$ 92,60
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
tamaño de rosca
20-400
Junta
joint: ST/NS24/29
Descripción general
These unique adapters greatly simplify the transfer of solids contained in round-bottom flasks into vials. Precision machined, chemically inert PTFE adapters will not seize in the joint. A wide range of thread sizes are available to accommodate most sample vials including scintillation vials (22 mm threads).
  • Transfers samples without exposure to air or moisture
  • Reduces sample losses due to air currents and static charge that can normally cause light solids to float or blow away when transferred open to the air
  • Excellent for transferring fluffy lyophilized samples, especially peptides
  • Transfers any freely flowing solid and eliminates weighing paper or other intermediate devices
Easy to Use:
Screw sample vial into bottle thread at top of adapter. Insert other end of adapter into flask joint. Invert assembly and gently tap vial on a soft surface to transfer solids from flask into sample vial.
Otras notas
Care must be used when tapping vial to prevent accidental breakage. Tapping on a cork ring or other soft surface is recommended.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)