Documentos clave
Z281360
Laboratory brushes
Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
envase
pkg of 4 ea
Longitud
30 in.
brush diam. × L
1/2 in. × 3 1/4 in.
Descripción general
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)