Documentos clave
Z260193
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 0.4-2.0 mL
Seleccione un Tamaño
1 EA
US$ 782,00
US$ 782,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 782,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 782,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
subdivision 0.1 mL
fabricante / nombre comercial
Fortuna Graf 101.107-27
intervalo de medición del volumen
0.4-2.0 mL
volumen
0.4-2 mL
Adaptador
GL 40
GL 45
Descripción general
For dispensing strong acids and aggressive chemicals
Features:
  • PTFE encased dispensing piston with sealing lip at its end
  • FIX adapter made of PTFE
  • Valve block made of PTFE, no metal springs in the valve - no corrosion
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
  • Ergonmic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
Supplied as Optifix® Safety dispenser, 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Información legal
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)