Documentos clave
Z214612
Micro photochemical reactor UV lamp
output5.5 watts
About This Item
Código UNSPSC:
41103817
Características
CE compliant: no
CSA approved: no
fabricante / nombre comercial
Ace Glass 1213208
disponibilidad
not available in EU, Canada
salida
5.5 watts
Descripción general
Cold cathode, low pressure, mercury arc, gaseous discharge lamps made of double bore quartz. Lamp power consumption is 5.5 watts, with principle output at 254 nanometers. Lamps are rated for 5000 hours of operation.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)